Learn More
STEMCELL Technologies TeSR™-E6, Size: 1 Kit
SDP
Catalog No. NC1724641
Encompass
TeSR™-E6 is a defined, serum- and xeno-free medium that is based on the formulation of TeSR™-E8™, but does not contain transforming growth factor β (TGF-β) or basic fibroblast growth factor (bFGF). It may be used as a basal medium for differentiation of human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells, or other applications where removal of the above cytokines is desirable.
Catalog No. NC1724641 Supplier STEMCELL Technologies Supplier No. 05946
TeSR™-E6 is a defined, serum- and xeno-free medium that is based on the formulation of TeSR™-E8™, but does not contain transforming growth factor β (TGF-β) or basic fibroblast growth factor (bFGF). It may be used as a basal medium for differentiation of human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells, or other applications where removal of the above cytokines is desirable.