Learn More
STEMCELL Technologies TeSR™-E7™ Medium for Reprogramming (2-Component), Size: 500 mL
SDP
Catalog No. NC1724639
Encompass
TeSR™-E7™ (2-Component) is an animal component-free and defined reprogramming culture medium optimized for the generation of human iPS cells without the use of feeders. It is based on the E7 formulation published by the laboratory of Dr. James Thomson (University of Wisconsin-Madison).
Catalog No. NC1724639 Supplier STEMCELL Technologies Supplier No. 05914
TeSR™-E7™ (2-Component) is an animal component-free and defined reprogramming culture medium optimized for the generation of human iPS cells without the use of feeders. It is based on the E7 formulation published by the laboratory of Dr. James Thomson (University of Wisconsin-Madison).