Learn More
STEMCELL Technologies TeSR™-E8™ Kit for hESC/hiPSC Maintenance, Size: 1 Kit
SDP
Catalog No. NC1424669
Encompass
TeSR™-E8™ is a feeder-free, animal component-free culture medium for human embryonic stem (ES) cells and human induced pluripotent stem (iPS) cells. It is based on the E8 formulation developed by the laboratory of Dr. James Thomson (University of Wisconsin-Madison), the lead research group behind the design of mTeSR™1, the most widely published feeder-free culture medium for pluripotent stem cells.Like mTeSR™1, TeSR™-E8™ medium is made with the highest level of quality and care. It contains only the essential components required for maintenance of ES and iPS cells, providing a simpler medium for the culture of pluripotent stem cells. TeSR™-E8™ may be used with either Corning Matrigel hESC-Qualified Matrix (Catalog #07181) or, for a completely defined xeno-free system, Vitronectin XF™ (Catalog #07180, a matrix developed and manufactured by Nucleus Biologics) as the culture matrix.
This item is not returnable. View return policy
Catalog No. NC1424669 Supplier STEMCELL Technologies Supplier No. 05990
TeSR™-E8™ is a feeder-free, animal component-free culture medium for human embryonic stem (ES) cells and human induced pluripotent stem (iPS) cells. It is based on the E8 formulation developed by the laboratory of Dr. James Thomson (University of Wisconsin-Madison), the lead research group behind the design of mTeSR™1, the most widely published feeder-free culture medium for pluripotent stem cells.Like mTeSR™1, TeSR™-E8™ medium is made with the highest level of quality and care. It contains only the essential components required for maintenance of ES and iPS cells, providing a simpler medium for the culture of pluripotent stem cells. TeSR™-E8™ may be used with either Corning Matrigel hESC-Qualified Matrix (Catalog #07181) or, for a completely defined xeno-free system, Vitronectin XF™ (Catalog #07180, a matrix developed and manufactured by Nucleus Biologics) as the culture matrix.