Learn More
MSC NES Test Indicator Contact Point, Carbide, 0.04 in., 1 ea.
Catalog No. 5000510964
Encompass
This carbide contact point is designed for precision measurement applications using lever-type dial indicators. It is engineered for durability and consistent performance in demanding metrology environments.
  • Carbide material for wear resistance
  • Length of 0.04 in.
  • Compatible with specified test indicators
Catalog No. 50-005-10964 Supplier MSC Supplier No. 01747047
This carbide contact point is designed for precision measurement applications using lever-type dial indicators. It is engineered for durability and consistent performance in demanding metrology environments.
  • Carbide material for wear resistance
  • Length of 0.04 in.
  • Compatible with specified test indicators