Learn More
MSC Mitutoyo Test Indicator Contact Point, Carbide Ball, M1.7 Thread, 11.2 mm Length, 1 ea
Catalog No. 5000510933
Encompass
These replacement contact points are designed for use with current test indicators requiring M1.7 threads. Manufactured in the USA, they feature a durable carbide ball tip to ensure reliable and precise measurements.
  • Carbide ball construction for enhanced durability
  • Compatible with current indicators featuring M1.7 threads
  • Engineered for precise measuring performance
Catalog No. 50-005-10933 Supplier MSC Supplier No. 01747294
These replacement contact points are designed for use with current test indicators requiring M1.7 threads. Manufactured in the USA, they feature a durable carbide ball tip to ensure reliable and precise measurements.
  • Carbide ball construction for enhanced durability
  • Compatible with current indicators featuring M1.7 threads
  • Engineered for precise measuring performance