Learn More
Med Vet International Blood Glucose Test Strips OneTouch Ultra Blue 50 Strips per Pack
Catalog No. 503043972
Encompass_Preferred
DoubleSure technology automatically checks each blood sample twice FastDraw capillary action draws sample in less than 1 second Provide visual confirmation of adequate blood sample
Catalog No. 50-304-3972 Supplier Med Vet International Supplier No. 850708BX
DoubleSure technology automatically checks each blood sample twice FastDraw capillary action draws sample in less than 1 second Provide visual confirmation of adequate blood sample