Learn More
Petroleum Analyzer Corporation Pour Point Test Tube complies with requirements of ASTM D5950
SDP
Catalog No. NC9687618
Encompass
Pour Point Test Tube complies with requirements of ASTM D5950 & D97
Catalog No. NC9687618 Supplier Petroleum Analyzer Corporation Supplier No. M00428
Pour Point Test Tube complies with requirements of ASTM D5950 & D97