Learn More
Hach Company pH Test Kit, 4.0 - 10.0 pH, Model 17N
Supplier:  Hach Company 147011
Encompass
Most natural waters range from pH 4 to pH 9, but commonly are above pH 7 because of carbonates and bicarbonates (alkalinity). The pH of water used in industry, boiler feed water, and swimming pools usually is kept within a narrow range. Simply add indicator to the sample and compare the color to the spots on the color disc to measure pH.
Catalog No. 50-199-4582