Learn More
Hach Company Hardness (Total) Test Kit, Model 5-EP MG-L
Supplier:  Hach Company 145401
Encompass
Total Hardness drop count titration/EDTA test kit with range of 20-400 mg/L; contains 100 tests.
Catalog No. 50-199-4566