Learn More
Hach Company Dissolved Oxygen AccuVac Kit
Supplier:  Hach Company 2515050
Encompass
Low DO levels usually indicate serious pollution. Adequate amounts are crucial for fish life, but conversely, DO must be excluded from boiler feed water to prevent corrosion.
Catalog No. 50-199-6164