Learn More
Hach Company Deluxe Water Conditioning Demonstration Test Kit, Model AD-16E
Catalog No. 501994586 Shop All Hach Company Products
Encompass
Designed to boost your testing capabilities, Hach's most complete water conditioning test kit includes tests and demonstrations for critical water quality parameters - soap consumption, softener, iron (0-7 mg/L), pH (5.6-8.4) and nitrate (0-40 mg/L), TDS (0-1990 ppm).
Catalog No. 50-199-4586 Supplier Hach Company Supplier No. 148410
Designed to boost your testing capabilities, Hach's most complete water conditioning test kit includes tests and demonstrations for critical water quality parameters - soap consumption, softener, iron (0-7 mg/L), pH (5.6-8.4) and nitrate (0-40 mg/L), TDS (0-1990 ppm).