Learn More
Hach Company Combined Ecology Test Kit, Model AL-36B
Catalog No. 501994604 Shop All Hach Company Products
Encompass
Utilizing drop count titration and color disc colorimetry, the Model AL-36B Test Kit is designed to meet the special needs of fish and wildlife conservation personnel by testing acidity, alkalinity, carbon dioxide, dissolved oxygen, hardness, and pH
Catalog No. 50-199-4604 Supplier Hach Company Supplier No. 180202
Utilizing drop count titration and color disc colorimetry, the Model AL-36B Test Kit is designed to meet the special needs of fish and wildlife conservation personnel by testing acidity, alkalinity, carbon dioxide, dissolved oxygen, hardness, and pH