Learn More
Hach Company General Boiler Water Test Kit, Model AL-94 MG-L, Drop Count Titration
Catalog No. 501994730 Shop All Hach Company Products
Encompass
Proper maintenance and economy of operation of modern boilers and cooling towers require a careful balance between water quality parameters and treatment chemicals. Used for controlling boiler and cooling water treatement and checking corrosion, this test kit is an economical solution for the small boiler operator who needs to monitor alkalinity (5-400 mg/L), chloride (5-400 mg/L), hardness (0-2,500 mg/L), iron (0-4 mg/L), and pH (4-10).
Catalog No. 50-199-4730 Supplier Hach Company Supplier No. 222403
Proper maintenance and economy of operation of modern boilers and cooling towers require a careful balance between water quality parameters and treatment chemicals. Used for controlling boiler and cooling water treatement and checking corrosion, this test kit is an economical solution for the small boiler operator who needs to monitor alkalinity (5-400 mg/L), chloride (5-400 mg/L), hardness (0-2,500 mg/L), iron (0-4 mg/L), and pH (4-10).