Learn More
Hach Company Alkalinity Test Kit, Model AL-AP, mg/L
Supplier:  Hach Company 2444301
Encompass
Caused by the presence of carbonates, bicarbonates, hydroxides, and other dissolved salts, alkalinity is important to food and beverage processors, boiler operators, and aquatic biologists. Kit utilizes drop count titration method and contains 100 tests.
Catalog No. 50-199-6008