Learn More
Hach Company Alkalinity Test Kit, Model AL-TA
Supplier:  Hach Company 2314500
Encompass
Caused by the presence of carbonates, bicarbonates, hydroxides, and other dissolved salts, alkalinity is important to food and beverage processors, boiler operators, and aquatic biologists. Utilizing drop count titration/sulfuric acid to determine alkalinity, this kit contains 100 tests.
Catalog No. 50-199-5780