Learn More
Hach Company Arsenic Low Range Test Kit
Supplier:  Hach Company 2800000
Encompass
Hach arsenic test strips provide results within a range of 0 - 500 ppb. This reliable method recovers 100% of inorganic arsenic in samples and allows detection of arsenic levels in the field to assess compliance with the EPA Arsenic Rule.
Catalog No. 50-199-6693