Learn More
Hach Company Ascorbic Acid Test Kit, Model ASC-1
Supplier:  Hach Company 2308100
Encompass
Model ASC-1 Ascorbic Acid Test Kit was developed in cooperation with beverage producers and uses the drop-count titration method to measure ascorbic acid (vitamin C). Utilizes drop count titration with a range of 10-200 mg/L. Contains approximately 100 tests.
Catalog No. 50-199-5768