Learn More
Med Vet International Comply Sterilization Bowie-Dick Test Pack Steam, 00135Lf, 6/Bag
Catalog No. 502831001
Encompass_Preferred
Designed to monitor the performance of vacuum-assisted 270° – 273° F steam sterilizers each day An early warning sheet that detects potential issues before they appear on the test sheet Early warning feature of the Comply Bowie-Dick Plus Test Pack permits scheduled sterilizer maintenance and helps to prevent unexpected failures Easy-to-read test sheet clearly identifies failures and can be filed for record keeping Lead free ink chemistry
Catalog No. 50-283-1001 Supplier Med Vet International Supplier No. 724429BG
 Restricted Item
Add to Cart
Add to Cart
Designed to monitor the performance of vacuum-assisted 270° – 273° F steam sterilizers each day An early warning sheet that detects potential issues before they appear on the test sheet Early warning feature of the Comply Bowie-Dick Plus Test Pack permits scheduled sterilizer maintenance and helps to prevent unexpected failures Easy-to-read test sheet clearly identifies failures and can be filed for record keeping Lead free ink chemistry