Learn More
Hach Company Boiler Feed and Scale Test Kit, Model BSC-1, Drop Count Titration
Catalog No. 501995851 Shop All Hach Company Products
Encompass
Proper maintenance and economy of operation of modern boilers and cooling towers require a careful balance between water quality parameters and treatment chemicals. An economical test kit for monitoring scale formation in small industrial boilers, the BSC-1 Boiler Feed and Scale Test Kit uses drop count titration methods to determine levels of alkalinity, chloride, and hardness.
Catalog No. 50-199-5851 Supplier Hach Company Supplier No. 2350300
Proper maintenance and economy of operation of modern boilers and cooling towers require a careful balance between water quality parameters and treatment chemicals. An economical test kit for monitoring scale formation in small industrial boilers, the BSC-1 Boiler Feed and Scale Test Kit uses drop count titration methods to determine levels of alkalinity, chloride, and hardness.