Learn More
Hach Company Carbon Dioxide Test Kit, Model CA-23
Supplier:  Hach Company 143601
Encompass
Carbon dioxide test kit with drop count titration covers ranges of 1.25-25, 2-40, and 5-100 mg/L as CO2, with increments of 1.25, 2, and 5 respectively. Includes rugged carrying case, prepackaged reagent set, detailed instructions, and all apparatus you need for fast, easy, and accurate carbon dioxide test results in the field.
Catalog No. 50-199-4559