Learn More
Hach Company Carbon Dioxide Test Kit, Model CA-DT
Supplier:  Hach Company 2064100
Encompass
Utilizing the Hach Digital Titrator, the Model CA-DT kit is used to determine carbon dioxide concentrations. Kit includes Digital Titrator, reagents and labware, manual and carrying case. Range: 10.1-1,000 mg/L. Contains approximately 50 tests.
Catalog No. 50-199-5208