Learn More
Hach Company Chloride Test Kit, Model CD-51
Supplier:  Hach Company 208601
Encompass
Found in nearly all natural waters, chloride affects human taste above 250 mg/L. High levels inhibit plant growth, and many industrial processes need to limit chloride concentration. The model CD-DT, multi-range kit, is supplied with the demineralizer bottle for high-range sample dilution. Contains 100 tests.
Catalog No. 50-199-4676