Learn More
Hach Company Chloride Test Kit, Model CD-DT
Catalog No. 501995199 Shop All Hach Company Products
Encompass
The single-parameter Model CD-DT uses the Hach Digital Titrator with Mercuric Nitrate Titrant Cartridges to determine chloride concentrations. Kit includes Digital Titrator, reagents and labware, manual and carrying case. Range: 10-8,000 mg/L. Contains approximately 100 tests.
Catalog No. 50-199-5199 Supplier Hach Company Supplier No. 2063500
The single-parameter Model CD-DT uses the Hach Digital Titrator with Mercuric Nitrate Titrant Cartridges to determine chloride concentrations. Kit includes Digital Titrator, reagents and labware, manual and carrying case. Range: 10-8,000 mg/L. Contains approximately 100 tests.