Learn More
Hach Company Chloride Test Kit, Model CDS-DT
Supplier: Hach Company 2580600
The single-parameter Model CDS-DT uses the Hach Digital Titrator with Silver Nitrate Titrant Cartridges to determine chloride concentrations. Kit includes Digital Titrator, reagents and labware, manual and carrying case. Range: 10-10,000 mg/L. Contains approximately 100 tests.