Learn More
Hach Company Chromium Test Kit, Model CH-12, Color Disc, 0.1-1.3 mg/L Cr
Supplier:  Hach Company 222800
Encompass
Widely used as a corrosion inhibitor in cooling towers and engine cooling systems, chromium is also used in surface finishing, leather tanning, and other industries. The most widely used form, hexavalent chromium, is very toxic and strict limits are placed on discharges.Method/Chemistry: Color disc. Range (mg/L): 0-1.3. Smallest Increment (mg/L): 0.1
Catalog No. 50-199-4733