Learn More
Hach Company Chlorine (Total) Test Kit, Model CN-21P
Catalog No. 501996010 Shop All Hach Company Products
Encompass
Chlorine Test Kits are manufactured by Hach Company to ensure compliance with stringent quality control standards. Using the Drop Count Titration method, the CN-21P Chlorine Test Kit is used to determine high concentrations of chlorine in sanitizing applications. 10-200 mg/L; contains 100 tests.*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. 50-199-6010 Supplier Hach Company Supplier No. 2444400
Chlorine Test Kits are manufactured by Hach Company to ensure compliance with stringent quality control standards. Using the Drop Count Titration method, the CN-21P Chlorine Test Kit is used to determine high concentrations of chlorine in sanitizing applications. 10-200 mg/L; contains 100 tests.*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.