Learn More
Hach Company Total Chlorine Color Disc Test Kit, Model CN-66T
Catalog No. 501994737 Shop All Hach Company Products
Encompass
One of Hach's most popular test kits, it uses powder DPD reagent that reacts with chlorine more quickly than tablet-form DPD, giving more accurate results. Powder DPD also has a considerable advantage over orthotolidine, a hazardous substance sometimes used as a chlorine test reagent. The simple, low-cost color disc method is the most accurate visual test kit method.*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. 50-199-4737 Supplier Hach Company Supplier No. 223103
One of Hach's most popular test kits, it uses powder DPD reagent that reacts with chlorine more quickly than tablet-form DPD, giving more accurate results. Powder DPD also has a considerable advantage over orthotolidine, a hazardous substance sometimes used as a chlorine test reagent. The simple, low-cost color disc method is the most accurate visual test kit method.*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.