Learn More
Hach Company Chlorine (Free & Total) and pH Test Kit, Model CN-67
Catalog No. 501994930 Shop All Hach Company Products
Encompass
Control of chlorine and pH in pools and spas to maintain safe bathing conditions and to protect against system corrosion. Supplied with color discs and comparator for monitoring chlorine and pH, the Free & Total Chlorine and pH Test Kit provides more accuracy than test kits using color cubes.
Catalog No. 50-199-4930 Supplier Hach Company Supplier No. 1411100
Control of chlorine and pH in pools and spas to maintain safe bathing conditions and to protect against system corrosion. Supplied with color discs and comparator for monitoring chlorine and pH, the Free & Total Chlorine and pH Test Kit provides more accuracy than test kits using color cubes.