Learn More
Hach Company Total Chlorine Test Kit, Model CN-DT, 20-2000 mg/L
Supplier:  Hach Company 2471100
Encompass
The most widely used disinfectant for drinking water, chlorine is also important for sanitizing swimming pools, cooling towers, other industrial equipment, and in the treatment of municipal wastewater. Its measurement and control are vital for both safety and economic reasons. Kit utilizes the Hach Digital Titrator and contains 100 tests.*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. 50-199-6049