Learn More
Med Vet International Respiratory Test Kit, Advin, COVID-19, Antigen Test, 66-9990-2, 25 Tests
Catalog No. 503044760
Encompass_Preferred
The Advin COVID-19 Antigen Test @Home is For FDA Emergency Use Authorization (EUA) only: https://www.fda.gov/medical-devices/covid-19-emergency-use-authorizations-medical-devices/in-vitro-diagnostics-euas-antigen-diagnostic-tests-sars-cov-2 The Advin COVID-19 Antigen Test @ Home is a lateral flow immunoassay device intended for the qualitative detection of nucleocapsid protein antigens from SARS-CoV-2 This test is authorized for non-prescription home use with self-collected anterior nasal (nares) swab samples from individuals aged 14 years or older or with adult-collected anterior nasal (nares) samples from individuals aged 2 years or older
Catalog No. 50-304-4760 Supplier Med Vet International Supplier No. 1236834KT
The Advin COVID-19 Antigen Test @Home is For FDA Emergency Use Authorization (EUA) only: https://www.fda.gov/medical-devices/covid-19-emergency-use-authorizations-medical-devices/in-vitro-diagnostics-euas-antigen-diagnostic-tests-sars-cov-2 The Advin COVID-19 Antigen Test @ Home is a lateral flow immunoassay device intended for the qualitative detection of nucleocapsid protein antigens from SARS-CoV-2 This test is authorized for non-prescription home use with self-collected anterior nasal (nares) swab samples from individuals aged 14 years or older or with adult-collected anterior nasal (nares) samples from individuals aged 2 years or older