Learn More
Hach Company Surface Water Test Kit
Supplier:  Hach Company 2559833
Encompass
By providing economical, easy-to-use tests for ammonia (0-2.4 mg/L), chlorine (0-3.4 mg/L), pH (0-14), nitrate (0-40 mg/L), dissolved oxygen (0.2-20 mg/L), phosphorus (0-40 mg/L), and temperature, this kit is an ideal tool for source water monitoring and wastewater treatment plants. The rugged, lightweight kit includes the convenient pH Pocket Pro Tester and all necessary reagents and apparatus for testing.
Catalog No. 50-199-6231