Learn More
Hach Company Glycol Visual Test Kit, Model EG-1
Supplier:  Hach Company 2186400
Encompass
Ethylene glycol (anti-freeze) contamination of automotive lubricants is an indication of a cracked engine block. Glycol presence also can indicate leaks in cooling systems. Test Kit Model EG-1 presents a purple color if ethylene glycol and other 1,2 glycols are present in oil or water. The kit contains approximately 25 tests and comes with a 14.4" x 8" x 7" Blue Polypropylene Carrying Case.
Catalog No. 50-199-5529