Learn More
Hach Company Sample Filtration and Degassing Kit
Supplier:  Hach Company 4397510
Encompass
For production of low-turbidity dilution water and removal of sample bubbles that interfere with turbidity measurement.
Catalog No. 50-199-7031