Learn More
Hach Company Glutaraldehyde Test Kit, Model GT-1
Supplier:  Hach Company 2587200
Encompass
From oil fields to cooling towers, hospitals to food plants, poultry farms to dental offices, Hach's Glutaraldehyde Test Kit can help monitor disinfection processes. The test kit measures from 0.5 to 2800 mg/L (ppm) glutaraldehyde.
Catalog No. 50-199-6273