Learn More
Hach Company Hardness (Total & Calcium) Test Kit, Model HA-4P
Supplier:  Hach Company 145700
Encompass
Hardness, Total & Calcium (as CaCO3) drop count titration/EDTA test kit, range 1-20 gpg; 100 tests.The kit contains all required reagents and apparatus in a rugged carrying case for analysis in the field.
Catalog No. 50-199-4568