Learn More
Hach Company Hardness (Total & Calcium) Test Kit, Model HA-4P MG-L
Supplier:  Hach Company 145701
Encompass
Hardness, Total & Calcium (Magnesium by difference) drop count titration/EDTA test kit, range 20 - 400 mg/L CaCO3, 100 tests.The Kit contains all required reagents and apparatus in a rugged carrying case for analysis in the field.
Catalog No. 50-199-4569