Learn More
Hach Company Hardness, Iron, and pH Test Kit, Model HA-62A
Supplier:  Hach Company 183701
Encompass
A multi-parameter test kit for hardness, iron (0-7 mg/L) & pH (6.6-8.4), this kit provides an easy-to-use, cost effective way to determine the quality of your water.
Catalog No. 50-199-4615