Learn More
Hach Company Hardness and Iron Color Disc Test Kit, Model HA-77
Supplier:  Hach Company 202300
Encompass
Hardness & Iron Water Quality Test Kit offers accuracy and portable utility in one package. Utilizing drop count titration (hardness) and color disc colorimetry (iron: 0-4 mg/L), this kit contains 100 tests.
Catalog No. 50-199-4660