Learn More
Hach Company Hardness, Iron, and pH Test Kit, Model HA-62B
Supplier:  Hach Company 183702
Encompass
Includes color disc and color comparator for colorimetric comparison of iron (0-7 mpg/L) and pH (4-10), and a drop-count titrator for hardness determination. Test kit produces a wide range of pH tests.
Catalog No. 50-199-4616