Learn More
Hach Company Hardness (Total & Calcium) Test Kit, Model HAC-DT
Supplier:  Hach Company 2063900
Encompass
Kit includes Digital Titrator, reagents and labware, manual and carrying case. Range: 10-4,000 mg/L. Contains approximately 100 of each (Total and Calcium) test.
Catalog No. 50-199-5206