Learn More
Hach Company Hydrazine Color Disc Test Kit, Model HY-2
Supplier:  Hach Company 184900
Encompass
Proper maintenance and economy of operation of modern boilers and cooling towers require a careful balance between water quality parameters and treatment chemicals. Utilizing the color disc method with a range of 0.02-1 mg/L, this kit contains 300 tests.
Catalog No. 50-199-4617