Learn More
Hach Company Hydrogen Peroxide Test Kit, Model HYP-1
Supplier:  Hach Company 2291700
Encompass
Model HYP-1 Hydrogen Peroxide Test Kit (as H₂O₂) utilizes drop count titration and contains approximately 100 tests.
Catalog No. 50-199-5743