Learn More
Hach Company Hypochlorite Test Kit, Model CN-HR
Supplier:  Hach Company 2687200
Encompass
Utilizing the simple, economical drop count titration method, this kit is used to determine high range hypochlorite solution concentrations (bleach solutions). Range: 5-15%, contains 100 tests.
Catalog No. 50-199-6546