Learn More
Hach Company Iron Color Disc Test Kit, Model IR-18
Supplier:  Hach Company 146400
Encompass
Using the simple color disc method, this kit measures Iron (medium range: 0.1 - 4 mg/L) and contains reagent for 100 tests.
Catalog No. 50-199-4573