Learn More
Hach Company Iron Color Disc Test Kit, Model IR-18A
Supplier:  Hach Company 146500
Encompass
Using the simple color disc method, this kit measures Iron (Range - 0.02-1 mg/L) and contains reagents for 100 tests.
Catalog No. 50-199-4575