Learn More
Hach Company Iron Color Disc Test Kit, Model IR-18B
Supplier:  Hach Company 146401
Encompass
Using the simple color disc method, this kit measures Iron (medium range - 0.2-7 mg/L) and contains reagent for 100 tests.
Catalog No. 50-199-4574