Learn More
Hach Company Iron (Ferrous) Color Disc Test Kit, Model IR-18C, 0.2-7 mg/L
Catalog No. 501996508 Shop All Hach Company Products
Encompass
The most accurate visual test kit method, Hach color discs provide fast, accurate comparisons. Range: 0.2-7 mg/L. Contains 100 tests.
Catalog No. 50-199-6508 Supplier Hach Company Supplier No. 2667200
The most accurate visual test kit method, Hach color discs provide fast, accurate comparisons. Range: 0.2-7 mg/L. Contains 100 tests.