Learn More
Hach Company Iron and pH Color Disc Test Kit, Model IR-23
Catalog No. 501994804 Shop All Hach Company Products
Encompass
Utilizing the simple, accurate color disc method, this Iron & pH Water Quality Test Kit provides accurate, on-site measurement of two important parameters - iron (0-7 mg/L) and pH (4-10) - to help maintain water quality within the desired range.
Catalog No. 50-199-4804 Supplier Hach Company Supplier No. 256200
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Utilizing the simple, accurate color disc method, this Iron & pH Water Quality Test Kit provides accurate, on-site measurement of two important parameters - iron (0-7 mg/L) and pH (4-10) - to help maintain water quality within the desired range.