Learn More
Hach Company Manganese Test Kit, Model MN-5
Supplier:  Hach Company 146700
Encompass
Manganese causes a bitter taste in water, and at concentrations above 0.1 mg/L, it causes objectionable stains on laundry and plumbing fixtures. Manganese control also is important in the beverage, paper, and textile industries, and in dye production and food processing. An economical solution, this kit uses the simple, accurate color disc method.
Catalog No. 50-199-4576