Learn More
Hach Company Manganese Color Disc Test Kit, Model MN-PAN
Supplier:  Hach Company 2350800
Encompass
Manganese causes a bitter taste in water, and at concentrations above 0.1 mg/L, it causes objectionable stains on laundry and plumbing fixtures. Manganese control also is important in the beverage, paper, and textile industries, and in dye production and food processing. Utilizing the simple color disc method, this kit measures manganese (Range: 0.05-0.7 mg/L) and contains 50 tests.
Catalog No. 50-199-5857